This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05260851_g1
See other VEGFB GE Assays ›
Gene Symbol:
Vegfb
Gene Name:
vascular endothelial growth factor B
Gene Aliases:
GW7_20680
Chromosome Location:
Chr.Un|NW_004624767: 22068107 - 22071491 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05260851_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vegfb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004852532.2 XP_004852589.2 6 - 7 958 75
XM_004852533.2 XP_004852590.2 6 - 7 857 75
XM_021264680.1 XP_021120339.1 7 - 8 1018 75
XM_021264681.1 XP_021120340.1 7 - 8 917 75
XM_021264682.1 XP_021120341.1 6 - 7 881 75
XM_021264683.1 XP_021120342.1 8 - 9 901 75
XM_021264684.1 XP_021120343.1 7 - 8 839 75
XM_021264685.1 XP_021120344.1 7 - 8 818 75
XM_021264686.1 XP_021120345.1 5 - 6 778 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0