Assay Details
Gene Symbol: Dnase1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021244463.1 | XP_021100122.1 | 6 - 7 | 691 | 68 | |
XM_021244464.1 | XP_021100123.1 | 5 - 6 | 574 | 68 | ||
XM_021244465.1 | XP_021100124.1 | 5 - 6 | 574 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
