Assay Details
Gene Symbol: LOC101725863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013078102.2 | XP_012933556.1 | 1 - 2 | 55 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: LOC101725863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013078102.2 | XP_012933556.1 | 1 - 2 | 55 | 63 |
Set Membership: |
![]() ![]() ![]() |