This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05247280_m1
See other TMBIM7P GE Assays ›
Gene Symbol:
Tmbim7p
Gene Name:
transmembrane BAX inhibitor motif containing 7, pseudogene
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624809: 746834 - 774582 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05247280_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmbim7p
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021242634.1 XP_021098293.1 5 - 6 1504 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0