Assay Details
Gene Symbol: Cabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852223.1 | XP_004852280.1 | 6 - 7 | 644 | 61 | |
XM_004852224.1 | XP_004852281.1 | 5 - 6 | 590 | 61 | ||
XM_004852225.1 | XP_004852282.1 | 5 - 6 | 485 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
