This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05243268_gH
See other BIRC3 GE Assays ›
Gene Symbol:
Birc3
Gene Name:
baculoviral IAP repeat containing 3
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624878: 523300 - 536062 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05243268_gH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Birc3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004870797.2 XP_004870854.1 2 - 3 2421 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

protease inhibitor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0