Assay Details
Gene Symbol: Pgbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847836.3 | XP_004847893.2 | 1 - 2 | 343 | 71 | |
XM_021260786.1 | XP_021116445.1 | 1 - 2 | 343 | 71 | ||
XM_021260789.1 | XP_021116448.1 | 1 - 2 | 343 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004847836.3, XM_021260786.1, XM_021260789.1 transcripts.
Set Membership: |
![]() ![]() |
