Assay Details
Gene Symbol: Ecm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004853978.3 | XP_004854035.1 | 5 - 6 | 546 | 62 | |
XM_021265871.1 | XP_021121530.1 | 4 - 5 | 432 | 62 | ||
XM_021265872.1 | XP_021121531.1 | 5 - 6 | 546 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
