Assay Details
Gene Symbol: Mea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013072825.2 | XP_012928279.1 | 2 - 3 | 386 | 66 | |
XM_021259793.1 | XP_021115452.1 | 2 - 3 | 406 | 66 | ||
XM_021259794.1 | XP_021115453.1 | 2 - 3 | 431 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
