This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05223110_m1
See other CREB3L1 GE Assays ›
Gene Symbol:
Creb3l1
Gene Name:
cAMP responsive element binding protein 3 like 1
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624767: 2215860 - 2247451 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05223110_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Creb3l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004851932.3 XP_004851989.1 3 - 4 837 52
XM_021264242.1 XP_021119901.1 3 - 4 838 52
XM_021264243.1 XP_021119902.1 3 - 4 837 52
XM_021264244.1 XP_021119903.1 3 - 4 514 52
XM_021264245.1 XP_021119904.1 3 - 4 736 52
XM_021264247.1 XP_021119906.1 3 - 4 837 52

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0