Assay Details
Gene Symbol: Adgrf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847139.2 | XP_004847196.2 | 7 - 8 | 2230 | 70 | |
XM_021259696.1 | XP_021115355.1 | 7 - 8 | 2230 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
