This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05219324_m1
See other GRK3 GE Assays ›
Gene Symbol:
Grk3
Gene Name:
G protein-coupled receptor kinase 3
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624747: 1575970 - 1758201 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05219324_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Grk3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004843328.3 XP_004843385.1 11 - 12 961 64
XM_021257130.1 XP_021112789.1 11 - 12 1011 64
XM_021257131.1 XP_021112790.1 11 - 12 1011 64
XM_021257132.1 XP_021112791.1 11 - 12 1011 64
XM_021257133.1 XP_021112792.1 11 - 12 1011 64
XM_021257134.1 XP_021112793.1 11 - 12 1011 64
XM_021257135.1 XP_021112794.1 9 - 10 752 64
XM_021257136.1 XP_021112795.1 11 - 12 1011 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0