This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05219160_m1
See other SUMF2 GE Assays ›
Gene Symbol:
Sumf2
Gene Name:
sulfatase modifying factor 2
Gene Aliases:
GW7_14815
Chromosome Location:
Chr.Un|NW_004624740: 8259140 - 8270194 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05219160_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sumf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004839975.3 XP_004840032.1 5 - 6 551 71
XM_013070460.2 XP_012925914.1 5 - 6 551 71
XM_013070462.2 XP_012925916.1 4 - 5 506 71
XM_021254316.1 XP_021109975.1 5 - 6 551 71
XM_021254317.1 XP_021109976.1 5 - 6 551 71
XM_021254318.1 XP_021109977.1 5 - 6 551 71
XM_021254319.1 XP_021109978.1 5 - 6 551 71
XM_021254320.1 XP_021109979.1 4 - 5 406 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0