Assay Details
Gene Symbol: Arhgap39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837666.3 | XP_004837723.1 | 8 - 9 | 2909 | 53 | |
XM_004837667.3 | XP_004837724.1 | 7 - 8 | 2816 | 53 | ||
XM_021250078.1 | XP_021105737.1 | 6 - 7 | 2281 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
