This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05207645_m1
See other UBE2D4 GE Assays ›
Gene Symbol:
Ube2d4
Gene Name:
ubiquitin conjugating enzyme E2 D4 (putative)
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624740: 7964827 - 7990611 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05207645_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2d4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004839938.3 XP_004839995.1 5 - 6 439 72
XM_004839939.3 XP_004839996.1 5 - 6 375 72
XM_021254303.1 XP_021109962.1 6 - 7 634 72
XM_021254304.1 XP_021109963.1 6 - 7 554 72
XM_021254307.1 XP_021109966.1 4 - 5 331 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0