This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05205903_m1
See other NR4A3 GE Assays ›
Gene Symbol:
Nr4a3
Gene Name:
nuclear receptor subfamily 4 group A member 3
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624825: 1101708 - 1141245 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05205903_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nr4a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004864935.2 XP_004864992.1 4 - 5 1795 64
XM_021244549.1 XP_021100208.1 6 - 7 1910 64
XM_021244550.1 XP_021100209.1 5 - 6 1867 64
XM_021244551.1 XP_021100210.1 5 - 6 1428 64
XM_021244552.1 XP_021100211.1 4 - 5 1817 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header