This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05203759_m1
See other GPR143 GE Assays ›
Gene Symbol:
Gpr143
Gene Name:
G protein-coupled receptor 143
Gene Aliases:
GW7_11425
Chromosome Location:
Chr.Un|NW_004624834: 7282031 - 7308920 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05203759_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr143
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021246375.1 XP_021102034.1 6 - 7 1209 101
XM_021246376.1 XP_021102035.1 7 - 8 1373 101
XM_021246377.1 XP_021102036.1 5 - 6 1117 101
XM_021246378.1 XP_021102037.1 7 - 8 1444 101
XM_021246379.1 XP_021102038.1 8 - 9 1190 101
XM_021246380.1 XP_021102039.1 7 - 8 1034 101
XM_021246381.1 XP_021102040.1 8 - 9 1017 101
XM_021246382.1 XP_021102041.1 7 - 8 1369 101
XM_021246383.1 XP_021102042.1 7 - 8 1373 101
XM_021246384.1 XP_021102043.1 7 - 8 1373 101
XM_021246385.1 XP_021102044.1 7 - 8 1371 101
XM_021246386.1 XP_021102045.1 6 - 7 1204 101
XM_021246387.1 XP_021102046.1 2 - 3 335 101

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021246387.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0