Assay Details
Gene Symbol: Mindy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013076140.2 | XP_012931594.1 | 1 - 2 | 228 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: Mindy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013076140.2 | XP_012931594.1 | 1 - 2 | 228 | 96 |
Set Membership: |
![]() ![]() ![]() |