Assay Details
Gene Symbol: Cabp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843412.3 | XP_004843469.1 | 2 - 3 | 275 | 77 | |
XM_004843413.3 | XP_004843470.2 | 2 - 3 | 275 | 77 | ||
XM_004843414.3 | XP_004843471.2 | 2 - 3 | 275 | 77 |
