Assay Details
Gene Symbol: Fkbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004872202.3 | XP_004872259.1 | 4 - 5 | 745 | 56 | |
XM_013068298.2 | XP_012923752.1 | 4 - 5 | 745 | 56 | ||
XM_021250152.1 | XP_021105811.1 | 4 - 5 | 747 | 56 | ||
XM_021250153.1 | XP_021105812.1 | 4 - 5 | 747 | 56 | ||
XM_021250154.1 | XP_021105813.1 | 4 - 5 | 752 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
