This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05186136_m1
See other LOC101723006 GE Assays ›
Gene Symbol:
LOC101723006
Gene Name:
putative RNA-binding protein Luc7-like 2
Gene Aliases:
Luc7l2
Chromosome Location:
Chr.Un|NW_004624765: 19768008 - 19851985 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05186136_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101723006
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004851264.3 XP_004851321.1 7 - 8 950 89
XM_004851266.3 XP_004851323.1 8 - 9 2208 89
XM_004851267.3 XP_004851324.1 8 - 9 846 89
XM_021263679.1 XP_021119338.1 10 - 11 1009 89
XM_021263681.1 XP_021119340.1 9 - 10 940 89
XM_021263682.1 XP_021119341.1 8 - 9 3251 89
XM_021263683.1 XP_021119342.1 8 - 9 1902 89
XM_021263684.1 XP_021119343.1 8 - 9 2014 89
XM_021263685.1 XP_021119344.1 7 - 8 733 89
XM_021263686.1 XP_021119345.1 9 - 10 975 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0