This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05184117_m1
See other POLG2 GE Assays ›
Gene Symbol:
Polg2
Gene Name:
DNA polymerase gamma 2, accessory subunit
Gene Aliases:
GW7_20874
Chromosome Location:
Chr.Un|NW_004624870: 179102 - 204474 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05184117_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polg2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004870230.3 XP_004870287.1 3 - 4 883 74
XM_021248640.1 XP_021104299.1 3 - 4 884 74
XM_021248641.1 XP_021104300.1 3 - 4 884 74
XM_021248642.1 XP_021104301.1 3 - 4 884 74
XM_021248643.1 XP_021104302.1 3 - 4 884 74
XM_021248644.1 XP_021104303.1 3 - 4 886 74
XM_021248645.1 XP_021104304.1 3 - 4 889 74
XM_021248646.1 XP_021104305.1 3 - 4 883 74
XM_021248647.1 XP_021104306.1 2 - 3 633 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header