This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05173062_m1
See other STK32B GE Assays ›
Gene Symbol:
Stk32b
Gene Name:
serine/threonine kinase 32B
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624755: 21992462 - 22294947 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05173062_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stk32b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004847305.3 XP_004847362.1 7 - 8 725 63
XM_013073058.2 XP_012928512.1 8 - 9 916 63
XM_013073059.2 XP_012928513.1 8 - 9 917 63
XM_013073061.2 XP_012928515.1 8 - 9 976 63
XM_013073062.2 XP_012928516.1 8 - 9 966 63
XM_013073063.2 XP_012928517.1 8 - 9 1169 63
XM_013073065.2 XP_012928519.1 8 - 9 1000 63
XM_013073066.1 XP_012928520.1 8 - 9 885 63
XM_021260306.1 XP_021115965.1 6 - 7 901 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header