This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05172705_m1
See other HCK GE Assays ›
Gene Symbol:
Hck
Gene Name:
HCK proto-oncogene, Src family tyrosine kinase
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624741: 455696 - 493821 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05172705_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hck
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004840586.2 XP_004840643.1 5 - 6 1026 63
XM_004840587.2 XP_004840644.1 5 - 6 1023 63
XM_021254896.1 XP_021110555.1 4 - 5 419 63
XM_021254897.1 XP_021110556.1 4 - 5 415 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0