Assay Details
Gene Symbol: LOC110344734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021239761.1 | XP_021095420.1 | 2 - 3 | 382 | 101 | |
XM_021239804.1 | XP_021095463.1 | 2 - 3 | 434 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021239761.1, XM_021239804.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
