This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05170703_g1
See other CITED1 GE Assays ›
Gene Symbol:
Cited1
Gene Name:
Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624903: 2596051 - 2601273 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05170703_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cited1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004872665.3 XP_004872722.1 2 - 3 406 79
XM_004872666.3 XP_004872723.1 2 - 3 863 79
XM_004872667.3 XP_004872724.1 2 - 3 410 79
XM_004872668.2 XP_004872725.1 2 - 3 433 79
XM_021250549.1 XP_021106208.1 2 - 3 176 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

transcription cofactor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0