This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05168203_m1
See other LOC101714324 GE Assays ›
Gene Symbol:
LOC101714324
Gene Name:
cat eye syndrome critical region protein 2
Gene Aliases:
Cecr2
Chromosome Location:
Chr.Un|NW_004624735: 9751621 - 9896266 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05168203_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101714324
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013068852.2 XP_012924306.1 7 - 8 932 77
XM_021249578.1 XP_021105237.1 7 - 8 932 77
XM_021249580.1 XP_021105239.1 7 - 8 933 77
XM_021249588.1 XP_021105247.1 7 - 8 931 77
XM_021249597.1 XP_021105256.1 7 - 8 931 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0