Assay Details
Gene Symbol: LOC101714324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068852.2 | XP_012924306.1 | 16 - 17 | 2918 | 68 | |
XM_021249580.1 | XP_021105239.1 | 16 - 17 | 2916 | 68 | ||
XM_021249582.1 | XP_021105241.1 | 15 - 16 | 2833 | 68 |

More Information
Set Membership: |
![]() ![]() |
