This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05160385_g1
See other CUNH1ORF109 GE Assays ›
Gene Symbol:
CUNH1orf109
Gene Name:
chromosome unknown C1orf109 homolog
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624764: 20409805 - 20418177 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05160385_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CUNH1orf109
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004850990.3 XP_004851047.1 1 - 2 327 85
XM_021263362.1 XP_021119021.1 1 - 2 332 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_004850990.3, XM_021263362.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0