Assay Details
Gene Symbol: LOC110343991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021236518.1 | XP_021092177.1 | 1 - 2 | 107 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: LOC110343991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021236518.1 | XP_021092177.1 | 1 - 2 | 107 | 77 |
Set Membership: |
![]() ![]() ![]() |