This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05153772_m1
See other CAB39L GE Assays ›
Gene Symbol:
Cab39l
Gene Name:
calcium binding protein 39 like
Gene Aliases:
GW7_05369
Chromosome Location:
Chr.Un|NW_004624748: 5238364 - 5408468 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05153772_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cab39l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004844158.3 XP_004844215.1 9 - 10 1141 72
XM_004844161.3 XP_004844218.1 7 - 8 1015 72
XM_021257834.1 XP_021113493.1 10 - 11 1249 72
XM_021257835.1 XP_021113494.1 11 - 12 1432 72
XM_021257836.1 XP_021113495.1 9 - 10 1313 72
XM_021257837.1 XP_021113496.1 8 - 9 1081 72
XM_021257838.1 XP_021113497.1 8 - 9 1039 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0