Assay Details
Gene Symbol: Fkbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852528.3 | XP_004852585.1 | 2 - 3 | 306 | 63 | |
XM_021264677.1 | XP_021120336.1 | 2 - 3 | 306 | 63 | ||
XM_021264678.1 | XP_021120337.1 | 3 - 4 | 261 | 63 | ||
XM_021264679.1 | XP_021120338.1 | 3 - 4 | 738 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
