Assay Details
Gene Symbol: Rpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004873993.2 | XP_004874050.1 | 8 - 9 | 701 | 89 | |
XM_004873994.2 | XP_004874051.1 | 8 - 9 | 718 | 89 | ||
XM_021251547.1 | XP_021107206.1 | 6 - 7 | 623 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
