Assay Details
Gene Symbol: Pifo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004853713.2 | XP_004853770.1 | 1 - 2 | 92 | 61 | |
XM_004853715.2 | XP_004853772.1 | 1 - 2 | 92 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004853713.2, XM_004853715.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
