This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05125343_g1
See other BOC GE Assays ›
Gene Symbol:
Boc
Gene Name:
BOC cell adhesion associated, oncogene regulated
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624731: 35256662 - 35330266 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05125343_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Boc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004835708.2 XP_004835765.1 8 - 9 1875 60
XM_004835709.2 XP_004835766.1 8 - 9 1879 60
XM_021263666.1 XP_021119325.1 9 - 10 2028 60
XM_021263670.1 XP_021119329.1 8 - 9 1555 60
XM_021263676.1 XP_021119335.1 8 - 9 1879 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0