This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05120912_m1
See other TIA1 GE Assays ›
Gene Symbol:
Tia1
Gene Name:
TIA1 cytotoxic granule associated RNA binding protein
Gene Aliases:
GW7_05839
Chromosome Location:
Chr.Un|NW_004624762: 3185647 - 3220410 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05120912_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tia1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004849911.3 XP_004849968.1 9 - 10 935 64
XM_004849913.3 XP_004849970.1 8 - 9 902 64
XM_013074072.2 XP_012929526.1 8 - 9 839 64
XM_021262493.1 XP_021118152.1 9 - 10 943 64
XM_021262494.1 XP_021118153.1 8 - 9 910 64
XM_021262496.1 XP_021118155.1 9 - 10 951 64
XM_021262497.1 XP_021118156.1 7 - 8 806 64
XM_021262498.1 XP_021118157.1 8 - 9 669 64
XM_021262499.1 XP_021118158.1 8 - 9 668 64
XM_021262500.1 XP_021118159.1 5 - 6 616 64
XM_021262501.1 XP_021118160.1 5 - 6 655 64
XM_021262502.1 XP_021118161.1 7 - 8 548 64
XM_021262503.1 XP_021118162.1 6 - 7 515 64
XM_021262504.1 XP_021118163.1 4 - 5 2571 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0