Assay Details
Gene Symbol: CUNH21orf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004842482.3 | XP_004842539.1 | 4 - 5 | 492 | 105 | |
XM_021256361.1 | XP_021112020.1 | 4 - 5 | 1867 | 105 | ||
XM_021256366.1 | XP_021112025.1 | 4 - 5 | 1235 | 105 |

More Information
Set Membership: |
![]() ![]() |
