This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05113987_g1
See other RHNO1 GE Assays ›
Gene Symbol:
Rhno1
Gene Name:
RAD9-HUS1-RAD1 interacting nuclear orphan 1
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624860: 222095 - 230708 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05113987_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rhno1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004869328.3 XP_004869385.1 2 - 3 292 89
XM_004869329.2 XP_004869386.1 2 - 3 296 89
XM_004869331.3 XP_004869388.1 3 - 4 634 89
XM_013067300.2 XP_012922754.1 3 - 4 487 89
XM_013067301.2 XP_012922755.1 3 - 4 3475 89
XM_021248057.1 XP_021103716.1 3 - 4 3512 89
XM_021248058.1 XP_021103717.1 3 - 4 3484 89
XM_021248059.1 XP_021103718.1 3 - 4 661 89
XM_021248060.1 XP_021103719.1 3 - 4 448 89
XM_021248061.1 XP_021103720.1 3 - 4 447 89
XM_021248062.1 XP_021103721.1 2 - 3 289 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0