This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05113753_m1
See other LYN GE Assays ›
Gene Symbol:
Lyn
Gene Name:
LYN proto-oncogene, Src family tyrosine kinase
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624886: 4030780 - 4168541 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05113753_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lyn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004871523.3 XP_004871580.1 8 - 9 1074 69
XM_004871524.3 XP_004871581.1 8 - 9 877 69
XM_004871525.3 XP_004871582.1 8 - 9 1011 69
XM_021249544.1 XP_021105203.1 8 - 9 809 69
XM_021249545.1 XP_021105204.1 8 - 9 1076 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0