Assay Details
Gene Symbol: Jam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004842220.2 | XP_004842277.1 | 5 - 6 | 1197 | 71 | |
XM_004842221.2 | XP_004842278.1 | 5 - 6 | 1200 | 71 | ||
XM_013071275.2 | XP_012926729.1 | 5 - 6 | 1218 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
