Assay Details
Gene Symbol: Sccpdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004853483.3 | XP_004853540.1 | 6 - 7 | 780 | 73 | |
XM_004853484.3 | XP_004853541.1 | 6 - 7 | 781 | 73 | ||
XM_021265500.1 | XP_021121159.1 | 6 - 7 | 783 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
