This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05093095_m1
See other GPRC5B GE Assays ›
Gene Symbol:
Gprc5b
Gene Name:
G protein-coupled receptor class C group 5 member B
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624782: 3166314 - 3184975 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05093095_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gprc5b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004855817.3 XP_004855874.1 3 - 4 1198 61
XM_013076302.2 XP_012931756.1 3 - 4 2217 61
XM_013076303.2 XP_012931757.1 3 - 4 2069 61
XM_013076304.2 XP_012931758.1 3 - 4 1142 61
XM_021237231.1 XP_021092890.1 2 - 3 2521 61
XM_021237232.1 XP_021092891.1 2 - 3 1053 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0