Assay Details
Gene Symbol: Ubl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004875011.2 | XP_004875068.1 | 2 - 3 | 239 | 104 | |
XM_004875012.2 | XP_004875069.1 | 2 - 3 | 239 | 104 | ||
XM_004875013.2 | XP_004875070.1 | 2 - 3 | 239 | 104 | ||
XM_004875014.2 | XP_004875071.1 | 2 - 3 | 239 | 104 | ||
XM_004875015.2 | XP_004875072.1 | 2 - 3 | 239 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
