Assay Details
Gene Symbol: Tm6sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852870.2 | XP_004852927.1 | 3 - 4 | 344 | 63 | |
XM_021264889.1 | XP_021120548.1 | 3 - 4 | 344 | 63 | ||
XM_021264890.1 | XP_021120549.1 | 3 - 4 | 344 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
