This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05081614_m1
See other H6PD GE Assays ›
Gene Symbol:
H6pd
Gene Name:
hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624818: 3029187 - 3052481 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05081614_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: H6pd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004863656.3 XP_004863713.1 2 - 3 1268 76
XM_004863661.3 XP_004863718.1 3 - 4 1978 76
XM_004863662.3 XP_004863719.1 2 - 3 1512 76
XM_004863663.2 XP_004863720.1 2 - 3 752 76
XM_004863664.3 XP_004863721.1 2 - 3 818 76
XM_013065128.2 XP_012920582.1 3 - 4 834 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0