Assay Details
Gene Symbol: Crybg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855440.3 | XP_004855497.1 | 10 - 11 | 4657 | 82 | |
XM_021236886.1 | XP_021092545.1 | 10 - 11 | 5064 | 82 | ||
XM_021236887.1 | XP_021092546.1 | 9 - 10 | 4655 | 82 | ||
XM_021236888.1 | XP_021092547.1 | 9 - 10 | 4731 | 82 |
