Assay Details
Gene Symbol: Crybg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855440.3 | XP_004855497.1 | 5 - 6 | 4078 | 95 | |
XM_021236886.1 | XP_021092545.1 | 5 - 6 | 4485 | 95 | ||
XM_021236887.1 | XP_021092546.1 | 4 - 5 | 4076 | 95 | ||
XM_021236888.1 | XP_021092547.1 | 4 - 5 | 4152 | 95 |
