This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05081197_m1
See other CRYBG1 GE Assays ›
Gene Symbol:
Crybg1
Gene Name:
crystallin beta-gamma domain containing 1
Gene Aliases:
Aim1
Chromosome Location:
Chr.Un|NW_004624780: 15709314 - 15952951 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05081197_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crybg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004855440.3 XP_004855497.1 3 - 4 1717 84
XM_021236886.1 XP_021092545.1 3 - 4 2124 84
XM_021236887.1 XP_021092546.1 2 - 3 1715 84
XM_021236888.1 XP_021092547.1 2 - 3 1791 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0