This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05080644_m1
See other NR1H4 GE Assays ›
Gene Symbol:
Nr1h4
Gene Name:
nuclear receptor subfamily 1 group H member 4
Gene Aliases:
GW7_15032
Chromosome Location:
Chr.Un|NW_004624750: 7221738 - 7303840 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05080644_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nr1h4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004844862.3 XP_004844919.3 4 - 5 1321 68
XM_004844863.3 XP_004844920.3 4 - 5 1333 68
XM_004844869.3 XP_004844926.1 6 - 7 1195 68
XM_013072272.2 XP_012927726.1 6 - 7 1209 68
XM_013072273.2 XP_012927727.1 7 - 8 1320 68
XM_013072274.2 XP_012927728.1 8 - 9 1401 68
XM_013072275.2 XP_012927729.1 7 - 8 1290 68
XM_013072276.2 XP_012927730.1 5 - 6 1068 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0