Assay Details
Gene Symbol: Lasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004859527.3 | XP_004859584.1 | 4 - 5 | 525 | 71 | |
XM_004859528.3 | XP_004859585.2 | 4 - 5 | 338 | 71 | ||
XM_004859529.3 | XP_004859586.2 | 4 - 5 | 332 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
